The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039675.1 | NP_001034764.1 | 21 - 22 | 3209 | 107 |
NM_001323619.1 | NP_001310548.1 | 18 - 19 | 2468 | 107 | |
NM_001323620.1 | NP_001310549.1 | 18 - 19 | 3006 | 107 | |
NM_001323621.1 | NP_001310550.1 | 18 - 19 | 2813 | 107 | |
NM_018671.4 | NP_061141.2 | 18 - 19 | 2858 | 107 | |
XM_005254963.3 | XP_005255020.1 | 18 - 19 | 2578 | 107 | |
XM_011521779.2 | XP_011520081.1 | 21 - 22 | 3454 | 107 | |
XM_011521780.2 | XP_011520082.1 | 17 - 18 | 2290 | 107 | |
XM_017022407.1 | XP_016877896.1 | 18 - 19 | 2508 | 107 | |
GenBank mRNA | AB014729.1 | - | 18 - 19 | 2400 | 107 |
AB014736.1 | - | 18 - 19 | 2442 | 107 | |
AK125523.1 | - | 7 - 8 | 1848 | 107 | |
AK125721.1 | - | 21 - 22 | 3209 | 107 | |
AK291622.1 | - | 18 - 19 | 2413 | 107 | |
AL357537.1 | - | 3 - 4 | 685 | 107 | |
AL357538.1 | - | 2 - 3 | 154 | 107 | |
BC006214.1 | - | 18 - 19 | 2441 | 107 | |
BC010995.2 | - | 5 - 6 | 448 | 107 | |
BC037992.1 | - | 18 - 19 | 2441 | 107 | |
BC045635.1 | - | 21 - 22 | 3209 | 107 |