The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202513.1 | NP_001189442.1 | 1 - 2 | 369 | 102 |
NM_001202514.1 | NP_001189443.1 | 1 - 2 | 369 | 102 | |
NM_002357.3 | NP_002348.1 | 1 - 2 | 369 | 102 | |
GenBank mRNA | AK301088.1 | - | 1 - 2 | 301 | 102 |
AK312734.1 | - | 1 - 2 | 302 | 102 | |
BC069377.1 | - | 1 - 2 | 163 | 102 | |
BC069433.1 | - | 1 - 2 | 163 | 102 | |
BC098396.1 | - | 1 - 2 | 339 | 102 | |
BC113531.1 | - | 1 - 2 | 222 | 102 | |
BC117260.1 | - | 1 - 2 | 222 | 102 | |
BC143831.1 | - | 1 - 2 | 222 | 102 | |
BC143832.1 | - | 1 - 2 | 222 | 102 | |
BC144601.1 | - | 1 - 2 | 222 | 102 | |
CR536495.1 | - | 1 - 2 | 79 | 102 | |
CR541692.1 | - | 1 - 2 | 79 | 102 | |
DB445256.1 | - | 1 - 2 | 365 | 102 | |
DC347823.1 | - | 1 - 2 | 370 | 102 | |
L06895.1 | - | 1 - 2 | 226 | 102 |