The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005415.4 | NP_005406.3 | 4 - 5 | 1102 | 64 |
XM_017004770.1 | XP_016860259.1 | 4 - 5 | 1073 | 64 | |
GenBank mRNA | AK312738.1 | - | 4 - 5 | 1021 | 64 |
BC019944.1 | - | 4 - 5 | 1009 | 64 | |
L20859.1 | - | 4 - 5 | 933 | 64 |