The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016474.4 | NP_057558.3 | 10 - 11 | 1196 | 70 |
NR_135523.1 | 9 - 10 | 1101 | 70 | ||
XM_017006554.1 | XP_016862043.1 | 8 - 9 | 1031 | 70 | |
GenBank mRNA | AF151046.1 | - | 8 - 9 | 934 | 70 |
AK091158.1 | - | 5 - 6 | 580 | 70 | |
BC013999.2 | - | 10 - 11 | 1172 | 70 | |
BC033897.1 | - | 10 - 11 | 1183 | 70 |