The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318781.1 | NP_001305710.1 | 14 - 15 | 2245 | 95 |
NM_182920.1 | NP_891550.1 | 15 - 16 | 2329 | 95 | |
GenBank mRNA | AB037733.1 | - | 14 - 15 | 1824 | 95 |
AF261918.1 | - | 15 - 16 | 2309 | 95 | |
AF488803.1 | - | 15 - 16 | 2329 | 95 | |
BC130578.1 | - | 15 - 16 | 2319 | 95 | |
BC171764.1 | - | 14 - 15 | 2235 | 95 |