The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318781.1 | NP_001305710.1 | 16 - 17 | 2501 | 114 |
NM_182920.1 | NP_891550.1 | 17 - 18 | 2585 | 114 | |
GenBank mRNA | AB037733.1 | - | 16 - 17 | 2080 | 114 |
AF261918.1 | - | 17 - 18 | 2565 | 114 | |
AF488803.1 | - | 17 - 18 | 2585 | 114 | |
BC130578.1 | - | 17 - 18 | 2575 | 114 | |
BC171764.1 | - | 16 - 17 | 2491 | 114 |