Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS9
Entrez Gene ID
56999
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
Gene Aliases
-
Chromosome Location
Chr.3: 64515654 - 64688000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318781.1 NP_001305710.1 1 - 2 145 103
NM_182920.1 NP_891550.1 1 - 2 145 103
GenBank mRNA AF488803.1 - 1 - 2 145 103
AK310571.1 - 1 - 2 456 103
BC130578.1 - 1 - 2 135 103
BC171764.1 - 1 - 2 135 103

View Full Product Details