Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFYVE19
Entrez Gene ID
84936
Gene Name
zinc finger FYVE-type containing 19
Gene Aliases
ANCHR, MPFYVE
Chromosome Location
Chr.15: 40807076 - 40814569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077268.1 NP_001070736.1 3 - 4 969 77
NM_001258420.1 NP_001245349.1 3 - 4 969 77
NM_001258421.1 NP_001245350.1 3 - 4 772 77
NM_032850.4 NP_116239.3 4 - 5 638 77
XM_017022683.1 XP_016878172.1 2 - 3 1614 77
XM_017022684.1 XP_016878173.1 3 - 4 956 77
GenBank mRNA AB209224.1 - - 2371 77
AF445414.1 - 4 - 5 480 77
AK027746.1 - 3 - 4 401 77
AK122779.1 - 3 - 4 759 77
AK225553.1 - 3 - 4 392 77
BC021092.1 - 3 - 4 266 77
DB012963.1 - 3 - 4 387 77

View Full Product Details