Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFYVE19
Entrez Gene ID
84936
Gene Name
zinc finger FYVE-type containing 19
Gene Aliases
ANCHR, MPFYVE
Chromosome Location
Chr.15: 40807076 - 40814569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077268.1 NP_001070736.1 9 - 10 1719 112
NM_001258420.1 NP_001245349.1 8 - 9 1515 112
NM_001258421.1 NP_001245350.1 9 - 10 1522 112
NM_032850.4 NP_116239.3 10 - 11 1388 112
XM_017022683.1 XP_016878172.1 8 - 9 2281 112
GenBank mRNA AB209224.1 - - 3312 112
AF445414.1 - 10 - 11 1230 112
AK027746.1 - 9 - 10 1151 112
AK122779.1 - 9 - 10 1509 112
AK225553.1 - 9 - 10 1142 112
BC021092.1 - 8 - 9 812 112
BI860679.1 - 3 - 4 205 112

View Full Product Details