Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF40
Entrez Gene ID
9810
Gene Name
ring finger protein 40
Gene Aliases
BRE1B, RBP95, STARING
Chromosome Location
Chr.16: 30761612 - 30800231 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207033.1 NP_001193962.1 6 - 7 1587 75
NM_001207034.1 NP_001193963.1 6 - 7 1587 75
NM_001286572.2 NP_001273501.1 6 - 7 948 75
NM_014771.3 NP_055586.1 6 - 7 1587 75
XM_011545997.1 XP_011544299.1 6 - 7 928 75
GenBank mRNA AB014561.1 - 6 - 7 859 75
AF122819.1 - 6 - 7 863 75
AK027406.1 - 6 - 7 877 75
AK075526.1 - 4 - 5 608 75
AK291666.1 - 6 - 7 877 75
BC006133.1 - 6 - 7 854 75
BC011769.2 - 6 - 7 854 75
BC018647.2 - 6 - 7 854 75
CR627431.1 - 6 - 7 916 75
KU178492.1 - 5 - 6 767 75
KU178493.1 - 5 - 6 767 75

View Full Product Details