Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF40
Entrez Gene ID
9810
Gene Name
ring finger protein 40
Gene Aliases
BRE1B, RBP95, STARING
Chromosome Location
Chr.16: 30761612 - 30800231 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207033.1 NP_001193962.1 9 - 10 1937 71
NM_001286572.2 NP_001273501.1 9 - 10 1298 71
NM_014771.3 NP_055586.1 9 - 10 1937 71
XM_011545997.1 XP_011544299.1 9 - 10 1278 71
GenBank mRNA AB014561.1 - 9 - 10 1209 71
AF122819.1 - 9 - 10 1213 71
BC006133.1 - 9 - 10 1204 71
BC011769.2 - 9 - 10 1204 71
BC018647.2 - 9 - 10 1204 71
BC030802.1 - 1 - 2 98 71
BX640763.1 - 6 - 7 571 71
CR627431.1 - 9 - 10 1266 71
KU178492.1 - 8 - 9 1117 71
KU178493.1 - 8 - 9 1117 71

View Full Product Details