The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168357.1 | NP_001161829.1 | 5 - 6 | 648 | 94 |
NM_005084.3 | NP_005075.3 | 5 - 6 | 739 | 94 | |
XM_005249408.4 | XP_005249465.1 | 5 - 6 | 723 | 94 | |
GenBank mRNA | AK290381.1 | - | 5 - 6 | 698 | 94 |
AK294471.1 | - | 4 - 5 | 521 | 94 | |
AK297066.1 | - | 2 - 3 | 315 | 94 | |
BC038452.1 | - | 5 - 6 | 648 | 94 | |
U20157.1 | - | 5 - 6 | 631 | 94 | |
U24577.1 | - | 5 - 6 | 686 | 94 |