Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOTCH4
Entrez Gene ID
4855
Gene Name
notch 4
Gene Aliases
INT3
Chromosome Location
Chr.6: 32194843 - 32224067 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004557.3 NP_004548.3 17 - 18 2826 59
NR_134949.1 18 - 19 2928 59
NR_134950.1 17 - 18 2826 59
GenBank mRNA BC140782.1 - 17 - 18 2781 59
BC144540.1 - 18 - 19 2889 59
BC144541.1 - 17 - 18 2781 59
U95299.1 - 17 - 18 2777 59

View Full Product Details