The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004557.3 | NP_004548.3 | 19 - 20 | 3258 | 98 |
NR_134949.1 | 20 - 21 | 3360 | 98 | ||
NR_134950.1 | 19 - 20 | 3258 | 98 | ||
GenBank mRNA | BC140782.1 | - | 19 - 20 | 3213 | 98 |
BC144540.1 | - | 20 - 21 | 3321 | 98 | |
BC144541.1 | - | 19 - 20 | 3213 | 98 | |
D63395.1 | - | 2 - 3 | 395 | 98 | |
U95299.1 | - | 19 - 20 | 3209 | 98 |