The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004557.3 | NP_004548.3 | 21 - 22 | 3889 | 68 |
GenBank mRNA | BC140782.1 | - | 21 - 22 | 3844 | 68 |
D63395.1 | - | 4 - 5 | 1026 | 68 | |
U95299.1 | - | 21 - 22 | 3840 | 68 |