The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004557.3 | NP_004548.3 | 28 - 29 | 5340 | 90 |
NR_134949.1 | 28 - 29 | 4909 | 90 | ||
NR_134950.1 | 27 - 28 | 4807 | 90 | ||
GenBank mRNA | AK131314.1 | - | 7 - 8 | 3670 | 90 |
AK293692.1 | - | 2 - 3 | 429 | 90 | |
BC140782.1 | - | 28 - 29 | 5295 | 90 | |
BC144540.1 | - | 28 - 29 | 4870 | 90 | |
BC144541.1 | - | 27 - 28 | 4762 | 90 | |
D63395.1 | - | 11 - 12 | 2477 | 90 | |
U95299.1 | - | 28 - 29 | 5291 | 90 |