Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOTCH4
Entrez Gene ID
4855
Gene Name
notch 4
Gene Aliases
INT3
Chromosome Location
Chr.6: 32194843 - 32224067 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004557.3 NP_004548.3 8 - 9 1651 73
NR_134950.1 8 - 9 1651 73
GenBank mRNA AK290785.1 - 8 - 9 1648 73
BC063815.1 - 8 - 9 1641 73
BC140782.1 - 8 - 9 1606 73
BC144541.1 - 8 - 9 1606 73
U95299.1 - 8 - 9 1602 73

View Full Product Details