Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIAS3
Entrez Gene ID
10401
Gene Name
protein inhibitor of activated STAT 3
Gene Aliases
ZMIZ5
Chromosome Location
Chr.1: 145848522 - 145859081 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006099.3 NP_006090.2 2 - 3 530 87
GenBank mRNA AB021868.1 - 2 - 3 437 87
AK027557.1 - 2 - 3 529 87
AK304039.1 - 2 - 3 530 87
BC001154.1 - 2 - 3 475 87
BC030556.1 - 2 - 3 486 87
BT007036.1 - 1 - 2 412 87
CR457090.1 - 1 - 2 412 87

View Full Product Details