Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIAS3
Entrez Gene ID
10401
Gene Name
protein inhibitor of activated STAT 3
Gene Aliases
ZMIZ5
Chromosome Location
Chr.1: 145848522 - 145859081 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006099.3 NP_006090.2 6 - 7 894 92
GenBank mRNA AB021868.1 - 6 - 7 801 92
AK027557.1 - 6 - 7 893 92
BC001154.1 - 6 - 7 839 92
BC030556.1 - 6 - 7 850 92
BT007036.1 - 5 - 6 776 92
CR457090.1 - 5 - 6 776 92

View Full Product Details