Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2Q2
Entrez Gene ID
92912
Gene Name
ubiquitin conjugating enzyme E2 Q2
Gene Aliases
-
Chromosome Location
Chr.15: 75843286 - 75901047 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005254787.1 XP_005254844.1 9 - 10 1042 97
XM_006720770.1 XP_006720833.1 6 - 7 805 97
XM_017022726.1 XP_016878215.1 9 - 10 1146 97
XM_017022727.1 XP_016878216.1 8 - 9 965 97
GenBank mRNA BC006827.1 - 7 - 8 545 97

View Full Product Details