Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLLT6
Entrez Gene ID
4302
Gene Name
MLLT6, PHD finger domain containing
Gene Aliases
AF17
Chromosome Location
Chr.17: 38705620 - 38729803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005937.3 NP_005928.2 10 - 11 1667 98
XM_006721911.2 XP_006721974.1 10 - 11 1589 98
XM_017024648.1 XP_016880137.1 5 - 6 1192 98
XM_017024649.1 XP_016880138.1 10 - 11 1667 98
XM_017024650.1 XP_016880139.1 10 - 11 1667 98
GenBank mRNA AB209633.1 - 10 - 11 1642 98
U07932.1 - 10 - 11 1654 98

View Full Product Details