Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLLT6
Entrez Gene ID
4302
Gene Name
MLLT6, PHD finger domain containing
Gene Aliases
AF17
Chromosome Location
Chr.17: 38705620 - 38729803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005937.3 NP_005928.2 4 - 5 362 81
XM_006721911.2 XP_006721974.1 4 - 5 362 81
XM_017024649.1 XP_016880138.1 4 - 5 362 81
XM_017024650.1 XP_016880139.1 4 - 5 362 81
GenBank mRNA AB209633.1 - 4 - 5 337 81
AK310435.1 - 4 - 5 362 81
AK310437.1 - 4 - 5 362 81
BC064612.1 - 4 - 5 450 81
U07932.1 - 4 - 5 349 81

View Full Product Details