Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLLT6
Entrez Gene ID
4302
Gene Name
MLLT6, PHD finger domain containing
Gene Aliases
AF17
Chromosome Location
Chr.17: 38705620 - 38729803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005937.3 NP_005928.2 16 - 17 2453 79
XM_006721911.2 XP_006721974.1 16 - 17 2375 79
XM_017024648.1 XP_016880137.1 11 - 12 1978 79
GenBank mRNA AB209633.1 - 16 - 17 2428 79
AK027133.1 - 5 - 6 571 79
AL133659.1 - 4 - 5 1432 79
BC007237.1 - 8 - 9 1404 79
U07932.1 - 16 - 17 2440 79

View Full Product Details