The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128.5 | NP_001119.3 | 6 - 7 | 949 | 87 |
GenBank mRNA | AB015317.1 | - | 6 - 7 | 654 | 87 |
AK128078.1 | - | 9 - 10 | 1504 | 87 | |
AK294546.1 | - | 7 - 8 | 802 | 87 | |
AK314484.1 | - | 6 - 7 | 954 | 87 |