The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016418.5 | NP_057502.2 | 15 - 16 | 2186 | 67 |
NM_181825.2 | NP_861546.1 | 15 - 16 | 2186 | 67 | |
NM_181828.2 | NP_861966.1 | 14 - 15 | 2060 | 67 | |
NM_181829.2 | NP_861967.1 | 14 - 15 | 2063 | 67 | |
NM_181830.2 | NP_861968.1 | 13 - 14 | 1937 | 67 | |
NM_181831.2 | NP_861969.1 | 13 - 14 | 1937 | 67 | |
NM_181832.2 | NP_861970.1 | 15 - 16 | 2186 | 67 | |
XM_017028810.1 | XP_016884299.1 | 15 - 16 | 1745 | 67 | |
GenBank mRNA | AF369658.1 | - | 15 - 16 | 2168 | 67 |
AF369663.1 | - | 14 - 15 | 1617 | 67 | |
AF369664.1 | - | 14 - 15 | 1620 | 67 | |
AF369665.1 | - | 13 - 14 | 1494 | 67 | |
AF369666.1 | - | 16 - 17 | 1859 | 67 | |
AF369667.1 | - | 15 - 16 | 1768 | 67 | |
AF369668.1 | - | 15 - 16 | 1743 | 67 | |
AF369700.1 | - | 7 - 8 | 810 | 67 | |
AF369701.1 | - | 15 - 16 | 2109 | 67 | |
BC003112.1 | - | 13 - 14 | 1766 | 67 |