The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000268.3 | NP_000259.1 | 4 - 5 | 890 | 72 |
NM_016418.5 | NP_057502.2 | 4 - 5 | 890 | 72 | |
NM_181825.2 | NP_861546.1 | 4 - 5 | 890 | 72 | |
NM_181828.2 | NP_861966.1 | 3 - 4 | 764 | 72 | |
NM_181829.2 | NP_861967.1 | 3 - 4 | 767 | 72 | |
NM_181830.2 | NP_861968.1 | 2 - 3 | 641 | 72 | |
NM_181831.2 | NP_861969.1 | 2 - 3 | 641 | 72 | |
NM_181832.2 | NP_861970.1 | 4 - 5 | 890 | 72 | |
XM_017028809.1 | XP_016884298.1 | 4 - 5 | 453 | 72 | |
XM_017028810.1 | XP_016884299.1 | 4 - 5 | 449 | 72 | |
GenBank mRNA | AF123570.1 | - | 4 - 5 | 497 | 72 |
AF369657.1 | - | 4 - 5 | 872 | 72 | |
AF369658.1 | - | 4 - 5 | 872 | 72 | |
AF369661.1 | - | 4 - 5 | 447 | 72 | |
AF369662.1 | - | 4 - 5 | 447 | 72 | |
AF369663.1 | - | 3 - 4 | 321 | 72 | |
AF369664.1 | - | 3 - 4 | 324 | 72 | |
AF369665.1 | - | 2 - 3 | 198 | 72 | |
AF369666.1 | - | 5 - 6 | 563 | 72 | |
AF369667.1 | - | 4 - 5 | 472 | 72 | |
AF369668.1 | - | 4 - 5 | 447 | 72 | |
AF369701.1 | - | 4 - 5 | 813 | 72 | |
AK297116.1 | - | 4 - 5 | 829 | 72 | |
BC003112.1 | - | 2 - 3 | 470 | 72 | |
BC020257.1 | - | 4 - 5 | 785 | 72 | |
CR456530.1 | - | 4 - 5 | 509 | 72 | |
L11353.1 | - | 4 - 5 | 666 | 72 | |
Z22664.1 | - | 4 - 5 | 621 | 72 |