The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000268.3 | NP_000259.1 | 1 - 2 | 557 | 73 |
NM_016418.5 | NP_057502.2 | 1 - 2 | 557 | 73 | |
NM_181825.2 | NP_861546.1 | 1 - 2 | 557 | 73 | |
NM_181829.2 | NP_861967.1 | 1 - 2 | 557 | 73 | |
NM_181832.2 | NP_861970.1 | 1 - 2 | 557 | 73 | |
NM_181833.2 | NP_861971.1 | 1 - 2 | 557 | 73 | |
GenBank mRNA | AF122827.1 | - | 1 - 2 | 164 | 73 |
AF123570.1 | - | 1 - 2 | 164 | 73 | |
AF369657.1 | - | 1 - 2 | 539 | 73 | |
AF369658.1 | - | 1 - 2 | 539 | 73 | |
AF369661.1 | - | 1 - 2 | 114 | 73 | |
AF369662.1 | - | 1 - 2 | 114 | 73 | |
AF369664.1 | - | 1 - 2 | 114 | 73 | |
AF369667.1 | - | 1 - 2 | 114 | 73 | |
AF369668.1 | - | 1 - 2 | 114 | 73 | |
AF369701.1 | - | 1 - 2 | 480 | 73 | |
AK297116.1 | - | 1 - 2 | 496 | 73 | |
BC020257.1 | - | 1 - 2 | 452 | 73 | |
CF138178.1 | - | 1 - 2 | 338 | 73 | |
CR456530.1 | - | 1 - 2 | 176 | 73 | |
L11353.1 | - | 1 - 2 | 333 | 73 | |
Z22664.1 | - | 1 - 2 | 288 | 73 |