The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080437.1 | NP_001073906.1 | 20 - 21 | 2710 | 83 |
XM_011510931.2 | XP_011509233.1 | 21 - 22 | 3024 | 83 | |
XM_011510932.2 | XP_011509234.1 | 21 - 22 | 3026 | 83 | |
XM_011510933.2 | XP_011509235.1 | 21 - 22 | 3030 | 83 | |
XM_011510934.2 | XP_011509236.1 | 21 - 22 | 3030 | 83 | |
XM_017003773.1 | XP_016859262.1 | 21 - 22 | 3026 | 83 | |
XM_017003774.1 | XP_016859263.1 | 20 - 21 | 2795 | 83 | |
XM_017003775.1 | XP_016859264.1 | 21 - 22 | 3028 | 83 | |
XM_017003776.1 | XP_016859265.1 | 20 - 21 | 2912 | 83 | |
XM_017003777.1 | XP_016859266.1 | 19 - 20 | 2803 | 83 | |
XM_017003778.1 | XP_016859267.1 | 20 - 21 | 2790 | 83 | |
XM_017003779.1 | XP_016859268.1 | 21 - 22 | 2726 | 83 | |
XM_017003780.1 | XP_016859269.1 | 20 - 21 | 2582 | 83 | |
XM_017003781.1 | XP_016859270.1 | 20 - 21 | 2572 | 83 | |
XM_017003782.1 | XP_016859271.1 | 16 - 17 | 1921 | 83 | |
GenBank mRNA | AF439717.1 | - | 14 - 15 | 1613 | 83 |
AK074062.1 | - | 19 - 20 | 2318 | 83 | |
AK074075.1 | - | 2 - 3 | 220 | 83 | |
BC027939.1 | - | 3 - 4 | 548 | 83 |