The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080437.1 | NP_001073906.1 | 16 - 17 | 2252 | 69 |
XM_011510931.2 | XP_011509233.1 | 17 - 18 | 2566 | 69 | |
XM_011510932.2 | XP_011509234.1 | 17 - 18 | 2568 | 69 | |
XM_011510933.2 | XP_011509235.1 | 17 - 18 | 2572 | 69 | |
XM_011510934.2 | XP_011509236.1 | 17 - 18 | 2572 | 69 | |
XM_011510937.2 | XP_011509239.1 | 17 - 18 | 2571 | 69 | |
XM_017003773.1 | XP_016859262.1 | 17 - 18 | 2568 | 69 | |
XM_017003774.1 | XP_016859263.1 | 16 - 17 | 2337 | 69 | |
XM_017003775.1 | XP_016859264.1 | 17 - 18 | 2570 | 69 | |
XM_017003777.1 | XP_016859266.1 | 15 - 16 | 2345 | 69 | |
XM_017003778.1 | XP_016859267.1 | 16 - 17 | 2332 | 69 | |
XM_017003779.1 | XP_016859268.1 | 17 - 18 | 2268 | 69 | |
XM_017003780.1 | XP_016859269.1 | 16 - 17 | 2124 | 69 | |
XM_017003781.1 | XP_016859270.1 | 16 - 17 | 2114 | 69 | |
XM_017003782.1 | XP_016859271.1 | 12 - 13 | 1463 | 69 | |
GenBank mRNA | AF439717.1 | - | 10 - 11 | 1155 | 69 |
AK074062.1 | - | 15 - 16 | 1860 | 69 |