Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDGFB
Entrez Gene ID
5155
Gene Name
platelet derived growth factor subunit B
Gene Aliases
IBGC5, PDGF-2, PDGF2, SIS, SSV, c-sis
Chromosome Location
Chr.22: 39223359 - 39245055 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002608.3 NP_002599.1 4 - 5 1551 91
NM_033016.3 NP_148937.1 4 - 5 451 91
GenBank mRNA BC029822.1 - 4 - 5 771 91
BC077725.1 - 4 - 5 1283 91
CR456538.1 - 4 - 5 467 91
CR541807.1 - 4 - 5 459 91
M12783.1 - 4 - 5 1551 91
X02744.1 - 4 - 5 576 91
X02811.1 - 4 - 5 1441 91
X03702.1 - 3 - 4 384 91
X83705.1 - 4 - 5 451 91

View Full Product Details