The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018046.4 | NP_060516.2 | 7 - 8 | 1690 | 95 |
GenBank mRNA | AK001145.1 | - | 2 - 3 | 202 | 95 |
AK289394.1 | - | 3 - 4 | 542 | 95 | |
AY500994.1 | - | 7 - 8 | 1609 | 95 | |
BC015478.1 | - | 2 - 3 | 441 | 95 | |
BC032844.1 | - | 7 - 8 | 1676 | 95 | |
BX648120.1 | - | 3 - 4 | 499 | 95 |