Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFP64
Entrez Gene ID
55734
Gene Name
ZFP64 zinc finger protein
Gene Aliases
ZNF338
Chromosome Location
Chr.20: 52084011 - 52191985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319146.1 NP_001306075.1 2 - 3 505 63
NM_199427.2 NP_955459.2 6 - 7 1264 63
XM_006723822.1 XP_006723885.1 2 - 3 434 63
XM_017027945.1 XP_016883434.1 6 - 7 1050 63
XM_017027947.1 XP_016883436.1 5 - 6 906 63
GenBank mRNA AK022690.1 - 2 - 3 505 63
AK222712.1 - 6 - 7 1058 63
BC021087.1 - 6 - 7 985 63
BT009760.1 - 6 - 7 915 63
DA731353.1 - 2 - 3 507 63

View Full Product Details