The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319146.1 | NP_001306075.1 | 3 - 4 | 566 | 65 |
NM_199427.2 | NP_955459.2 | 7 - 8 | 1325 | 65 | |
XM_006723822.1 | XP_006723885.1 | 3 - 4 | 495 | 65 | |
XM_017027945.1 | XP_016883434.1 | 7 - 8 | 1111 | 65 | |
XM_017027947.1 | XP_016883436.1 | 6 - 7 | 967 | 65 | |
GenBank mRNA | AK022690.1 | - | 3 - 4 | 566 | 65 |
AK222712.1 | - | 7 - 8 | 1119 | 65 | |
BC021087.1 | - | 7 - 8 | 1046 | 65 | |
BT009760.1 | - | 7 - 8 | 976 | 65 | |
DA731353.1 | - | 3 - 4 | 568 | 65 |