Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RALGAPB
Entrez Gene ID
57148
Gene Name
Ral GTPase activating protein non-catalytic beta subunit
Gene Aliases
KIAA1219, RalGAPbeta
Chromosome Location
Chr.20: 38472806 - 38578861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282917.1 NP_001269846.1 17 - 18 2719 69
NM_001282918.1 NP_001269847.1 17 - 18 2849 69
NM_020336.3 NP_065069.1 17 - 18 2861 69
XM_005260462.2 XP_005260519.1 18 - 19 2855 69
XM_005260463.2 XP_005260520.1 18 - 19 2855 69
XM_005260464.2 XP_005260521.1 18 - 19 2843 69
XM_005260465.2 XP_005260522.1 17 - 18 2819 69
XM_017027966.1 XP_016883455.1 18 - 19 2843 69
XM_017027967.1 XP_016883456.1 17 - 18 2807 69
XM_017027968.1 XP_016883457.1 17 - 18 2381 69
XM_017027969.1 XP_016883458.1 18 - 19 2872 69
XM_017027970.1 XP_016883459.1 2 - 3 343 69
GenBank mRNA AB033045.3 - 17 - 18 2839 69
AK123576.1 - 16 - 17 2520 69
AK125260.1 - 5 - 6 622 69
AK125916.1 - 12 - 13 1818 69
AL834436.1 - 15 - 16 2083 69
BC047482.1 - 17 - 18 2824 69
CR936800.1 - 17 - 18 2625 69

View Full Product Details