The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282917.1 | NP_001269846.1 | 4 - 5 | 711 | 74 |
NM_001282918.1 | NP_001269847.1 | 4 - 5 | 853 | 74 | |
NM_020336.3 | NP_065069.1 | 4 - 5 | 853 | 74 | |
XM_005260462.2 | XP_005260519.1 | 4 - 5 | 811 | 74 | |
XM_005260463.2 | XP_005260520.1 | 4 - 5 | 811 | 74 | |
XM_005260464.2 | XP_005260521.1 | 4 - 5 | 811 | 74 | |
XM_005260465.2 | XP_005260522.1 | 4 - 5 | 811 | 74 | |
XM_017027966.1 | XP_016883455.1 | 4 - 5 | 811 | 74 | |
XM_017027967.1 | XP_016883456.1 | 4 - 5 | 811 | 74 | |
XM_017027968.1 | XP_016883457.1 | 3 - 4 | 337 | 74 | |
XM_017027969.1 | XP_016883458.1 | 5 - 6 | 876 | 74 | |
GenBank mRNA | AB033045.3 | - | 4 - 5 | 843 | 74 |
AB621815.1 | - | 3 - 4 | 633 | 74 | |
AK123576.1 | - | 3 - 4 | 512 | 74 | |
AK304242.1 | - | 4 - 5 | 809 | 74 | |
AL834436.1 | - | 2 - 3 | 75 | 74 | |
BC047482.1 | - | 4 - 5 | 816 | 74 |