Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFN1
Entrez Gene ID
55669
Gene Name
mitofusin 1
Gene Aliases
hfzo1, hfzo2
Chromosome Location
Chr.3: 179347692 - 179394944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033540.2 NP_284941.2 11 - 12 1354 125
XM_005247596.3 XP_005247653.2 11 - 12 1312 125
XM_011512963.2 XP_011511265.1 8 - 9 940 125
GenBank mRNA AB043588.1 - 10 - 11 125
AB043589.1 - 11 - 12 125
AF054986.1 - 1 - 2 111 125
AF329637.1 - 10 - 11 1228 125
AK314306.1 - 11 - 12 1311 125
BC020661.1 - 11 - 12 1320 125
BC040557.1 - 11 - 12 1354 125
CR627379.1 - 8 - 9 1003 124
U95822.1 - 7 - 8 125

View Full Product Details