Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFN1
Entrez Gene ID
55669
Gene Name
mitofusin 1
Gene Aliases
hfzo1, hfzo2
Chromosome Location
Chr.3: 179347692 - 179394944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033540.2 NP_284941.2 4 - 5 533 101
XM_005247596.3 XP_005247653.2 4 - 5 491 101
GenBank mRNA AB043588.1 - 3 - 4 498 101
AB043589.1 - 4 - 5 498 101
AF329637.1 - 3 - 4 407 101
AK000700.1 - 4 - 5 490 101
AK314306.1 - 4 - 5 490 101
BC020661.1 - 4 - 5 499 101
BC040557.1 - 4 - 5 533 101
CR627379.1 - 1 - 2 183 101

View Full Product Details