The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003086.2 | NP_003077.2 | 11 - 12 | 1237 | 72 |
XM_005266096.2 | XP_005266153.1 | 12 - 13 | 1328 | 72 | |
XM_006717241.2 | XP_006717304.1 | 12 - 13 | 1312 | 72 | |
XM_006717242.3 | XP_006717305.1 | 12 - 13 | 2241 | 72 | |
XM_017015058.1 | XP_016870547.1 | 11 - 12 | 2757 | 72 | |
GenBank mRNA | AF032387.1 | - | 11 - 12 | 1603 | 72 |