Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAPC4
Entrez Gene ID
6621
Gene Name
small nuclear RNA activating complex polypeptide 4
Gene Aliases
PTFalpha, SNAP190
Chromosome Location
Chr.9: 136375571 - 136400220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003086.2 NP_003077.2 15 - 16 1991 100
XM_005266096.2 XP_005266153.1 16 - 17 2082 100
XM_006717241.2 XP_006717304.1 16 - 17 2066 100
XM_006717242.3 XP_006717305.1 16 - 17 2995 100
XM_006717244.2 XP_006717307.1 1 - 2 476 100
XM_017015058.1 XP_016870547.1 15 - 16 3511 100
GenBank mRNA AF032387.1 - 15 - 16 2357 100

View Full Product Details