Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNAPC4
Entrez Gene ID
6621
Gene Name
small nuclear RNA activating complex polypeptide 4
Gene Aliases
PTFalpha, SNAP190
Chromosome Location
Chr.9: 136375571 - 136400220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003086.2 NP_003077.2 18 - 19 2392 60
XM_005266096.2 XP_005266153.1 19 - 20 2483 60
XM_006717241.2 XP_006717304.1 19 - 20 2467 60
XM_006717242.3 XP_006717305.1 19 - 20 3396 60
XM_006717244.2 XP_006717307.1 4 - 5 834 60
XM_017015058.1 XP_016870547.1 18 - 19 3828 60
GenBank mRNA AF032387.1 - 18 - 19 2758 60

View Full Product Details