The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130064.1 | NP_001123536.1 | 3 - 4 | 1415 | 67 |
NM_002045.3 | NP_002036.1 | 2 - 3 | 1019 | 67 | |
XM_017006128.1 | XP_016861617.1 | 2 - 3 | 2664 | 67 | |
GenBank mRNA | AF279774.1 | - | 3 - 4 | 964 | 67 |
AK289699.1 | - | 3 - 4 | 1209 | 67 | |
AK290100.1 | - | 3 - 4 | 1088 | 67 | |
AK313658.1 | - | 2 - 3 | 716 | 67 | |
BC007936.2 | - | 2 - 3 | 676 | 67 | |
BT019771.1 | - | 2 - 3 | 633 | 67 | |
BT019810.1 | - | 2 - 3 | 633 | 67 | |
BX648458.1 | - | 3 - 4 | 1087 | 67 | |
M25667.1 | - | 2 - 3 | 724 | 67 |