The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001131018.1 | NP_001124490.1 | 3 - 4 | 448 | 117 |
XM_005251893.4 | XP_005251950.4 | 3 - 4 | 314 | 117 | |
XM_006717039.3 | XP_006717102.2 | 3 - 4 | 314 | 117 | |
XM_011518482.2 | XP_011516784.2 | 3 - 4 | 314 | 117 | |
XM_011518483.1 | XP_011516785.1 | 3 - 4 | 431 | 117 | |
GenBank mRNA | AF234161.1 | - | 2 - 3 | 138 | 117 |
AK027287.1 | - | 3 - 4 | 431 | 117 | |
BC021163.2 | - | 3 - 4 | 427 | 117 |