Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
Chromosome Location
Chr.9: 128166065 - 128204383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131017.1 NP_001124489.1 5 - 6 757 118
XM_005251892.3 XP_005251949.2 5 - 6 628 118
XM_006717039.3 XP_006717102.2 4 - 5 551 118
XM_011518481.1 XP_011516783.1 5 - 6 628 118
XM_017014595.1 XP_016870084.1 5 - 6 740 118
GenBank mRNA AF234161.1 - 3 - 4 375 118
AK303288.1 - 5 - 6 740 118

View Full Product Details