The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282802.1 | NP_001269731.1 | 4 - 5 | 537 | 71 |
NM_020654.4 | NP_065705.3 | 5 - 6 | 587 | 71 | |
XM_011513038.2 | XP_011511340.1 | 5 - 6 | 576 | 71 | |
XM_011513039.2 | XP_011511341.1 | 4 - 5 | 501 | 71 | |
XM_011513045.2 | XP_011511347.1 | 5 - 6 | 577 | 71 | |
XM_017006924.1 | XP_016862413.1 | 5 - 6 | 464 | 71 | |
XM_017006925.1 | XP_016862414.1 | 5 - 6 | 552 | 71 | |
XM_017006928.1 | XP_016862417.1 | 5 - 6 | 577 | 71 | |
GenBank mRNA | AF199458.1 | - | 4 - 5 | 535 | 71 |
AK297356.1 | - | - | 498 | 71 | |
BC062674.1 | - | 5 - 6 | 534 | 71 | |
BC129988.1 | - | 5 - 6 | 514 | 71 |