Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COQ2
Entrez Gene ID
27235
Gene Name
coenzyme Q2, polyprenyltransferase
Gene Aliases
CL640, COQ10D1, MSA1, PHB:PPT
Chromosome Location
Chr.4: 83138568 - 83285129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015697.7 NP_056512.5 6 - 7 1098 68
XM_017008031.1 XP_016863520.1 6 - 7 1221 68
GenBank mRNA AJ621061.3 - 6 - 7 1098 68
AK129583.1 - 6 - 7 854 68
BC008804.2 - 6 - 7 955 68
BC020728.1 - 6 - 7 995 68
BC116454.1 - 1 - 2 142 68
BQ363759.1 - 3 - 4 374 68

View Full Product Details