The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033504.2 | NP_277039.1 | 1 - 2 | 137 | 63 |
XM_005270408.3 | XP_005270465.1 | 1 - 2 | 220 | 63 | |
GenBank mRNA | AY027543.1 | - | 1 - 2 | 74 | 63 |
BC001418.3 | - | 1 - 2 | 137 | 63 | |
BC013953.2 | - | 1 - 2 | 137 | 63 | |
BC023260.1 | - | 1 - 2 | 174 | 63 |