The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005590.3 | NP_005581.2 | 13 - 14 | 1794 | 100 |
NM_005591.3 | NP_005582.1 | 13 - 14 | 1687 | 100 | |
XM_005274008.3 | XP_005274065.1 | 11 - 12 | 1402 | 100 | |
XM_006718842.3 | XP_006718905.1 | 13 - 14 | 1794 | 100 | |
XM_011542837.2 | XP_011541139.1 | 13 - 14 | 2216 | 100 | |
XM_017017772.1 | XP_016873261.1 | 13 - 14 | 1794 | 100 | |
XM_017017773.1 | XP_016873262.1 | 13 - 14 | 1687 | 100 | |
GenBank mRNA | AF022778.1 | - | 13 - 14 | 1668 | 100 |
AF073362.1 | - | 12 - 13 | 1498 | 100 | |
AK095388.1 | - | 13 - 14 | 1796 | 100 | |
AK308318.1 | - | 13 - 14 | 1685 | 99 | |
BC063458.1 | - | 12 - 13 | 1595 | 100 | |
U37359.1 | - | 13 - 14 | 1669 | 100 |