Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUL5
Entrez Gene ID
8065
Gene Name
cullin 5
Gene Aliases
VACM-1, VACM1
Chromosome Location
Chr.11: 108008682 - 108107766 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003478.3 NP_003469.2 5 - 6 1218 138
XM_005271682.2 XP_005271739.1 4 - 5 825 138
XM_011543013.1 XP_011541315.1 5 - 6 1002 138
XM_017018363.1 XP_016873852.1 5 - 6 1002 138
XM_017018364.1 XP_016873853.1 5 - 6 563 138
XM_017018365.1 XP_016873854.1 4 - 5 527 138
GenBank mRNA AF017061.1 - 5 - 6 554 138
AF327710.1 - 5 - 6 551 138
AK125294.1 - 5 - 6 1167 138
AK292575.1 - 5 - 6 981 138
BC063306.1 - 5 - 6 981 138
X81882.1 - 5 - 6 640 138

View Full Product Details