The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278503.1 | NP_001265432.1 | 3 - 4 | 336 | 103 |
NM_152713.4 | NP_689926.1 | 2 - 3 | 248 | 103 | |
XM_011542807.2 | XP_011541109.1 | 2 - 3 | 275 | 103 | |
GenBank mRNA | AK290040.1 | - | 2 - 3 | 198 | 103 |
AK308902.1 | - | 2 - 3 | 191 | 103 | |
BC020965.2 | - | 2 - 3 | 181 | 103 | |
BC048348.1 | - | 1 - 2 | 134 | 103 | |
BT007100.1 | - | 1 - 2 | 89 | 103 | |
BX649102.1 | - | 3 - 4 | 274 | 103 | |
DA577274.1 | - | 2 - 3 | 248 | 103 | |
L38961.1 | - | 2 - 3 | 195 | 103 | |
L47337.1 | - | 2 - 3 | 205 | 103 |