The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142505.1 | NP_001135977.1 | 2 - 3 | 341 | 91 |
NM_022169.4 | NP_071452.2 | 2 - 3 | 565 | 91 | |
GenBank mRNA | AJ300465.1 | - | 1 - 2 | 228 | 91 |
AJ308237.1 | - | 1 - 2 | 240 | 91 | |
AK289830.1 | - | 2 - 3 | 341 | 91 | |
AK290035.1 | - | 2 - 3 | 502 | 91 | |
BC041091.1 | - | 2 - 3 | 565 | 91 | |
DA241043.1 | - | 2 - 3 | 341 | 91 |